VistaLab Technologies™ Ovation™ Electronic Single Channel Pipette

Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions

Overview
Brand: VistaLab Technologies™

Manufacturer Part Number: 106505ML

UNSPSC: 41121507

Code: 50

Additional Details:
Additional Details: Weight: 0.40000kg



Product Code. 11952215

Quantity Price
1 £ 1110.06 / Each
Estimated Shipment
25-05-2017
Add to basket


Description and Specification

Specification

Color Gray
Warranty 1 Year
Display LCD
Compatible Tips VistaLab™ tip
Number of Speeds 5
Volume (Metric) 100 to 5000μL
Number of Channels 1
Increments 10μL
Finger Style Ergonomic
Model Right hand
For Use With (Equipment) VistaLab™ tip
Accuracy ±0.6%
Battery Type Lithium Ion
Documentation