VistaLab Technologies™ MLA™ Brand Pipet Tips: Nonsterile
For use with MLA Brand pipetters
Overview
Brand: VistaLab Technologies™
Manufacturer Part Number: 9048
X100 PIPETTOR TIP OVATION PIPETTORS BOX, NON-STERIle for volumes 100µL to 5000µL 5mL polypropylene
UNSPSC: 41121600
Code: 04
Additional Details:
Additional Details: Weight: 6.25000kg
Product Code. 11905722
Quantity Price
1 £ 32.33 / Pack of 100
Estimated Shipment
18-04-2017
Add to basket
Description and Specification
Specification
For Use With (Equipment) MLA Pipetters
Autoclavable Yes
Tip Style Macro
Packaging Dense
Volume (Metric) 5mL