missing translation for 'onlineSavingsMsg'
Learn More

VISTA Rat anti-Mouse, PE, Clone: MIH64, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen 12-1083-82

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943164

  • £226.00 / 100µg

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Rat Monoclonal Antibody

VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

VISTA
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9D659
B7H5; B7-H5; C10orf54; DD1alpha; Dies1; GI24; PD-1H; PD-H1; Platelet receptor Gi24; PP2135; SISP1; Sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; Vsir; V-type immunoglobulin domain-containing suppressor of T-cell activation
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
MIH64
PE
VSIR
Liquid
VSIR
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
74048
Videos
SDS
Documents