missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit
Shop All Macherey-Nagel Products
Click to view available options
Range:
0 to 0.20mg/L
0 to 0.50mg/L
No. of Tests:
100
300
For Use With (Application):
Iron determination
Manganese determination
Unit Size:
100 tests
Each
Description
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.
- Visual evaluation
- Environment-friendly, without toxic reagents
- Economically priced
- Convenient handling
- Highest accuracy due to extremely narrow gradation
- Highest sensitivity down to 0.002mg/L due to longer measuring tubes
- Reagent bottles with clear dosing instructions
- Compensation of turbidities and colors
- Refill packs available
Specifications
Specifications
| Analytes | Iron (Fe) |
| Color | Color change: Yellow to Red |
| Description | Highly sensitive |
| Format | HE Colorimetric Test Kit |
| For Use With (Application) | Iron determination |
| Graduations | 0.0 - 0.01 - 0.02 - 0.03 - 0.04 - 0.05 - 0.07 - 0.10 - 0.15 - 0.20mg/L Fe |
| Includes | Reagents and test equipment (e.g. glass tubes with screw caps, comparator block, color comparison disk) |
| No. of Tests | 300 |
| Quantity | 1 Each |
| Range | 0 to 0.20mg/L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction