Learn More
Macherey-Nagel™ Visocolor™ alpha Carbonate Hardness
Add reagent one drop at a time until the color changes at pH 4.5. Macherey-Nagel™ Visocolor™ alpha Carbonate Hardness performs titrimetric determination with hydrochloric acid against a mixed indicator. Each drop is scientifically equivalent to a specific measurement.
Brand: Macherey-Nagel™ 935016
This item is not returnable.
View return policy
Description
Visocolor™ alpha is the most simple titrimetric test kit, which is suitable for visual evaluation only. With multicomponent reagents, the test kits are limited in precision and accuracy but represent an inexpensive method for screening tests of non-turbid and uncolored water samples. The reagent bottles are packed in practical blister packs with the test instructions on the back.
- Carbonate hardness is calcium and magnesium ions that present as carbonate or hydrogen carbonate
- Reaction principle analogous to APHA 2320, DIN EN ISO 9963-1 C23 and -2 C24
- Used specimens can be flushed down the drain with tap water
Specifications
Titrimetric determination of carbonate hardness | |
Reagent, titration syringe | |
1 Each | |
Titration with hydrochloric acid against a mixed indicator |
Reaction principle analogous to APHA 2320, DIN EN ISO 9963-1 C23 & -2 C24 | |
100 | |
1 drop = 1 °d (corresp. to 18mg/L CaCO3) | |
Visocolor™ alpha Test Kit |