Learn More
Invitrogen™ Human NAMPT, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75758
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Visfatin (human) Recombinant Protein, RP-75758, from Invitrogen.
This product contains purified recombinant human Visfatin protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽60.2 kDa.Specifications
P43490 | |
Control, Western Blot | |
10135 | |
100μg | |
N/A | |
RUO | |
NAMPT | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
0.88mg/mL | |
tris HCl with 50% glycerol and no preservative | |
Human NAMPT | |
E. Coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human NAMPT | |
Human | |
Hu | |
Recombinant | |
Recombinant |