missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Viral Production Medium, prototype
Prototype, is a chemically defined, serum-free, protein-free medium specially developed for growth and transfection of Viral Production Cells 2.0
Brand: Gibco™ A49842BA
This item is not returnable.
View return policy
Description
- Gibco Viral Production Medium, prototype, is a chemically defined, serum-free, protein-free medium specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells) (Cat. No. A50532) for adeno-associated virus (AAV) production as part of the Gibco AAV-MAX Helper-Free AAV production system.
- Viral Production Medium, prototype, requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use.
Specifications
Store at 2–8°C, protected from light. | |
Mammalian Cells | |
Liquid | |
Bag | |
AAV-MAX | |
RUO |
HEK293 | |
Suspension Cell Culture | |
GlutaMAX | |
AAV Production | |
10 L |
For Research Use Only. Not for use in diagnostic procedures. This product is a prototype and performance characteristics of this product have not been established.