missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Viral haemorrhagic septicemia virus ELISA Kit

Enzyme-linked immunosorbent assay for the detection of viral haemorrhagic septicemia virus (VHSV).

Brand:  Abnova™ KA4894.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16019175

  • £334.56 / 1 set


Estimated Shipment
18-06-2020
Add to basket

Description

Description

  • Used for the in vitro quantitative determination of human SOD1 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-9
Specifications

Specifications

Colorimetric
Viral haemorrhagic septicemia virus ELISA Kit is an enzyme-linked immunosorbent assay for the detection of viral haemorrhagic septicemia virus (VHSV).
Microtitre Plate: 96 wells, Negative Antigen: 0.7mL, Positive Antigen: 0.7mL, RAVHSV: 6mL, Conjugate: 12mL, TMB-Complete 5: 15mL, Wash Solution: 20x concentrated buffer (20mL), Stop Solution: Acid solution ready to use (12mL)
For research use only (RUO)
Store the kit at 4°C. Do not freeze.
Homogenate
HRP
Qualitative
Viral haemorrhagic septicemia virus ELISA Kit
50 μL (Prediluted)
1 Kit
Videos
Documents