missing translation for 'onlineSavingsMsg'
Learn More

Viper™ Application Switching Solutions Kits

Order this single kit to get all the easy, zero-dead-volume fittings and capillaries for your application switching LC system.

Brand:  Thermo Scientific 6040.2806

UNSPSC : 41115720

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11382473

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Description

Conventional stainless steel HPLC connectors use a ferrule and a nut to establish a connection. A flaw of this design is that a void volume-free connection is not guaranteed, particularly when changing the tubing between differently shaped threads (e.g. when changing a column).
  • Viper connectors are standard for Dionex RSLC system connections
  • Compatible with virtually any third party UHPLC or HPLC instrument, valve, and column hardware
Description & Specifications

Specifications

UltiMate 3000 x2 Dual SD System
180µm
Various
Ultimate™ 3000 x2 Dual SD System
Automated/unattended fast method changes
1250bar/18125psi
Stainless Steel
Documentation