missing translation for 'onlineSavingsMsg'
Learn More

Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™, Acros Organics

£62.60 - £281.10

Chemical Identifiers

CAS 1826-67-1
Molecular Formula C2H3BrMg
Molecular Weight (g/mol) 131.25
InChI Key RMGJCSHZTFKPNO-UHFFFAOYSA-M
Synonym vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide
PubChem CID 74584
IUPAC Name magnesium;ethene;bromide
SMILES C=[CH-].[Mg+2].[Br-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13478598 View Documents Acros Organics
445971000
100mL £62.60
100mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-06-2020
Please to purchase this item. Need a web account? Register with us today!
13438608 View Documents Acros Organics
445978000
800mL £281.10
800mL View alternative products
Save Up to 
»
Estimated Shipment
24-06-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1826-67-1
131.25
vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide
magnesium;ethene;bromide
C2H3BrMg
RMGJCSHZTFKPNO-UHFFFAOYSA-M
74584
C=[CH-].[Mg+2].[Br-]

Specifications

Vinylmagnesium bromide
−11°C
0.965
96-47-9
CH2CHMgBr
1M solution in 2-MeTHF
RMGJCSHZTFKPNO-UHFFFAOYSA-M
magnesium;ethene;bromide
74584
0.9650g/mL
AcroSeal™ Glass bottle
100mL
C2H3BrMg
vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide
Solubility in water: reacts
C=[CH-].[Mg+2].[Br-]
131.25
131.25
Safety and Handling

Safety and Handling

Product Identifier
  • Vinylmagnesium bromide, 1M solution in 2-MeTHF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.