missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Vinylferrocene

Brand:  Alfa Aesar™ 017450.06

Code : NEW

Additional Details : CAS Number : 1271-51-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11383347

  • £263.40 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Vinylferrocene
1271-51-8
212.07
MFCD00001434

Specifications

Vinylferrocene
Powder
MFCD00001434
80° to 85°C (0.7mmHg)
62° (144°F)
1271-51-8
212.07
52° to 55°C
5g
Videos
SDS