missing translation for 'onlineSavingsMsg'
Learn More

Vinylbenzyl trimethylammonium chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3779892
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
26616-35-3
C12H18ClN
211.73
MFCD00080690
This item is not returnable. View return policy
26616-35-3
C12H18ClN
211.73
MFCD00080690

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 240°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH2CHC6H4CH2N(CH3)3Cl
Quantity 25 g
Formula Weight 211.73
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Vinylbenzyl trimethylammonium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'