missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinylacetic acid, tech., 90%, unstabilized

Catalog Number p-211883
61.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
625-38-7
C4H6O2
86.09
MFCD00002782
PVEOYINWKBTPIZ-UHFFFAOYSA-N
3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
32743
CHEBI:35897
but-3-enoic acid
OC(=O)CC=C
625-38-7
C4H6O2
86.09
MFCD00002782
PVEOYINWKBTPIZ-UHFFFAOYSA-N
3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
32743
CHEBI:35897
but-3-enoic acid
OC(=O)CC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -39°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 163°C
Flash Point 65°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula H2C=CHCH2CO2H
Refractive Index 1.4239 to 1.4259
Quantity 25 g
Beilstein 02, 407
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 86.09
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Vinylacetic acid, tech., 90%, unstabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'