missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Sulfone polystyrene resin, 1% DVB, 0.4-1.2mmol/g, 100-200 mesh, ACROS Organics™

Brand:  Acros Organics 358280010

UNSPSC : 12350000

Code : 29

Additional Details : Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10048751

  • £42.20 / 1g


Estimated Shipment
20-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Vinyl Sulfone polystyrene resin
1% DVB, 0.4-1.2 mmol/g, 100-200 mesh
MFCD03458981

Specifications

Beads
White to off-white
0.4 to 1.2mmol/g
Glass bottle
1% DVB, 0.4-1.2 mmol/g, 100-200 mesh
1g
crosslinking: 1% DVB
MFCD03458981
100 to 200 mesh
Vinyl Sulfone polystyrene resin
Documentation

Technical Documents