Allsigns International™ Vinyl Safety Sign Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA149/SA.

Safety sign WA149/SA hazard warning organicperoxide 200mm x 300mm self adhesive vinyl

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12335678

Quantity Price
1 £ 12.75 / Each
Estimated Shipment
30-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Used to Instruct, Advise and Forewarn Staff and Visitors of Potential Dangers Arising In and Around the Workplace
Width (Metric) 300mm
Adhesive Type Self
Quantity 1Each
Type Safety Sign Label
Resistant To Chemicals
Material Vinyl
Color Black Markings with Yellow on White Background
Message Organic Peroxide
Dimensions (L x W) 200 x 300mm
Shape Rectangle
Length (Metric) 200mm

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace. Color - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive

Documentation