Allsigns International™ Self-adhesive Vinyl Hazard Warning Signs

Warn workers and visitors about potential safety hazards by affixing Allsigns Interntional™ Self-adhesive Vinyl Hazard Warning Signs in relevant workplace areas and situations.

Overview
Brand: Allsigns International™

Manufacturer Part Number: WA158S

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12315698

Quantity Price
1 £ 12.64 / Each
Estimated Shipment
18-05-2017
Add to basket


Description and Specification

Specification

Color Black & yellow
Dimensions (L x W) 200mm x 300mm
Material Vinyl
Message Laser Beam
Shape Rectangle
Adhesive Type Self
Resistant To Chemicals

  • Black symbol and text, yellow background
  • Symbol - triangle
  • Self-adhesive
  • Display warning messages

Documentation