missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allsigns International Safety sign WA217/SA 200mm x 300mm hazard warning self adhesive vinyl
Brand: Allsigns International WA217S
Safety sign WA217/SA 200mm x 300mm hazard warning self adhesive vinyl
Code : 04
Additional Details : Weight : 0.01000kg