Allsigns International™ Vinyl Safety Sign Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA142/SA.

Safety sign WA142/SA hazard warning dangeroussubstances 200mm x 300mm self adhesive vinyl

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12315668

Quantity Price
1 £ 12.19 / Each
Estimated Shipment
02-03-2017
Add to basket


Description and Specification

Specification

Width (Metric) 300mm
Adhesive Type Self
Quantity 1Each
Type Safety Sign Label
Resistant To Chemicals
Color Black Markings with Yellow on White Background
Material Vinyl
Message Dangerous substances
Dimensions (L x W) 200 x 300mm
Shape Rectangle
Length (Metric) 200mm
Documentation

Technical documents