missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Self-adhesive Vinyl Hazard Warning Signs

Warn workers and visitors about potential safety hazards by affixing Allsigns Interntional™ Self-adhesive Vinyl Hazard Warning Signs in relevant workplace areas and situations.

Brand:  Allsigns International™ WA85S

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12355708

  • £12.67 / Each


Estimated Shipment
17-07-2017
Add to basket

Specifications

Description

  • Black symbol and text, yellow background
  • Symbol - triangle
  • Self-adhesive
  • Display warning messages
Description & Specifications

Specifications

Black & yellow
Vinyl
Rectangle
Chemicals
200mm x 300mm
Dangerous Chemicals
Self
Documentation

Technical Documents