missing translation for 'onlineSavingsMsg'
Learn More

ALLSIGNS International™ Vinyl Safety Sign

Instruct, advise and inform staff and visitors that an action must be carried out to secure a safer working environment with the Allsigns International™ Vinyl Safety Sign.

Brand:  ALLSIGNS International™ WA83S

UNSPSC : 55121700

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12375628

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Description

  • Self-adhesive
    • Blue symbol and background, white text
    • Symbol - circle
Description & Specifications

Specifications

200mm
Safety Sign
Vinyl
200 x 300mm
Trip Hazard
300mm
1/Ea.