missing translation for 'onlineSavingsMsg'
Learn More

ALLSIGNS International™ Vinyl Safety Sign

Instruct, advise and inform staff and visitors that an action must be carried out to secure a safer working environment with the Allsigns International™ Vinyl Safety Sign.

Brand:  ALLSIGNS International™ SA80S

UNSPSC : 55121718

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12319907

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Description

  • Self-adhesive
    • Blue symbol and background, white text
    • Symbol - circle
Description & Specifications

Specifications

300mm
Safety Sign
Vinyl
300 x 100mm
First Aid
100mm
1/Ea.
Documentation

Technical Documents