missing translation for 'onlineSavingsMsg'
Learn More

ALLSIGNS International™ Vinyl Safety Sign

Instruct, advise and inform staff and visitors that an action must be carried out to secure a safer working environment with the Allsigns International™ Vinyl Safety Sign.

Brand:  ALLSIGNS International™ SA89S

Code : 04

Additional Details : Weight : 0.01504kg

 View more versions of this product

Product Code. 12309887

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Self-adhesive
    • Blue symbol and background, white text
    • Symbol - circle
Specifications

Specifications

Vinyl
200 x 300mm
200mm
Safety Sign
1/Ea.
Self-adhesive
First Aid
300mm
Videos
Documentation

Technical Documents