Allsigns International™ Vinyl Mandatory Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: MA87S

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12359977

Quantity Price
1 £ 11.9 / Each
Estimated Shipment
05-06-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Type Mandatory Safety Sign
Color Blue symbol and background white text
For Use With (Application) These signs are designed to be used to indicate suitable Exit routes and Safe areas for disabled people
Material Vinyl
Dimensions (L x W) 200 x 300mm
Adhesive Type Self-adhesive
Message Ear protection must be worn in this area
Documentation

Technical documents