Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA89/SA.

Safety sign WA89/SA hazard warning danger guarddogs 200mm x 300mm self adhesive vinyl Allsigns,

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12395628

Quantity Price
1 £ 11.97 / Each
Estimated Shipment
30-03-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Danger guard dogs
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 200 x 300mm