Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA68/SA.

Safety sign WA68/SA hazard warning dangerasbestos 200mm x 300mm self adhesive vinyl

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12335628

Quantity Price
1 £ 11.97 / Each
Estimated Shipment
27-02-2017
Add to basket


Description and Specification

Specification

Dimensions (L x W) 200 x 300mm
Material Vinyl
Message Danger Asbestos
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive