Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA7A/SA.

Safety sign WA7A/SA hazard warning danger 110volts 200mm x 300mm self adhesive vinyl Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345598

Quantity Price
1 £ 11.97 / Each
Estimated Shipment
03-03-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Danger 110 volts
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents