Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA155S

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12355688

Quantity Price
1 £ 12.75 / Each
Estimated Shipment
05-06-2017
Add to basket


Description and Specification

Specification

Message Toxic Gas
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Material Vinyl
Quantity 1Each
For Use With (Application) To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace
Color Black Symbol and Text, Yellow Background
Dimensions (L x W) 200 x 300mm

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace.Colour - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive

Documentation