Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA148/SA.

Safety sign WA148/SA hazard warning spontaneouslycombustible 200mm x 300mm self adhesive vinyl

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12315678

Quantity Price
1 £ 12.14 / Each
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Spontaneously combustible
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents