missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Vinyl Hazard Warning Safety Sign

Brand:  Allsigns International™ EC33S

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12335708

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Specifications

200mm
Hazard Warning Safety Sign
Self-adhesive
1/Ea.
Risk of Freezing
300mm
Vinyl
Documentation

Technical Documents