missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Vinyl Hazard Warning Safety Sign

Brand:  Allsigns International™ WA41S

UNSPSC : 55121700

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12315618

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Specifications

200mm
Hazard Warning Safety Sign
Self-adhesive
1/Ea.
Mind the step
300mm
Vinyl
Documentation

Technical Documents