Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA135/SA.

Safety sign hazard warning caution wet paint selfadhesive vinyl 600mm x 450mm WA135/SA Allsigns,

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12355658

Quantity Price
1 £ 29.33 / Each
Estimated Shipment
30-03-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Hazard warning caution wet paint
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 600 x 450mm