missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Vinyl Hazard Warning Safety Sign

Brand:  Allsigns International™ WA153S

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12315688

  • £12.58 / Each


Estimated Shipment
14-08-2017
Add to basket

Specifications

Description

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace.Colour - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive
Description & Specifications

Specifications

200mm
Hazard Warning Safety Sign
Black symbol and text, Yellow background
1/Ea.
Explosive substance
300mm
Vinyl
Documentation