Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA26A/SA.

Safety sign WA26A/SA hazard warning danger ofdeath 200mm x 300mm self adhesive vinyl Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12335608

Quantity Price
1 £ 11.97 / Each
Estimated Shipment
03-04-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Danger of death
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents