Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA28/SA.

Safety sign WA28/SA hazard warning danger highvoltage 200mm x 300mm self adhesive vinyl

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12355608

Quantity Price
1 £ 11.74 / Each
Estimated Shipment
09-03-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Danger high voltage
Quantity 1Each
Color Black symbol and text, yellow background
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 200 x 300mm

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace.Colour - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive

Documentation