Allsigns International™ Vinyl Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA214/SA.

Safety sign WA214/SA hazard warning automaticdoor arrow pointing right 200mm x 75mm self

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12395638

Quantity Price
1 £ 9.91 / Each
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Message Automatic door right
Quantity 1Each
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive
Dimensions (L x W) 200 x 75mm