missing translation for 'onlineSavingsMsg'
Learn More

Vinyl chloroformate, 99%, stabilized, ACROS Organics™

Brand:  Acros Organics 275480100

Code : 29

Additional Details : CAS Number : 5130-24-5 Weight : 0.06250kg Transport : UN number : 2742 Chem class : 1a Pack group : II

Quantity 10mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10665083

  • £368.30 / 10mL


EU Stock 4
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Vinyl chloroformate
5130-24-5
99%
CH2=CHOC(O)Cl
MFCD00000641
99%
≥98.5%
C3H3ClO2
106.51

Specifications

Vinyl chloroformate
5130-24-5
Glass bottle
Liquid
99%
C3H3ClO2
106.51
03, 28
1.2500g/mL
10mL
99%
Authentic
1.4090 to 1.4110 (20°C, 589nm)
≥98.5%
50 to 100ppm BHT
CH2=CHOC(O)Cl
MFCD00000641
08,530
Clear colorless
Videos
SDS