missing translation for 'onlineSavingsMsg'
Learn More

Vinorelbine ditartrate, Tocris Bioscience™

Selective mitotic microtubule antagonist

Brand:  Tocris (Bio-Techne) 340150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462954

  • £507.02 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Selective mitotic microtubule antagonist that exhibits > 20 fold selectivity over axonal microtubules. Inhibits proliferation of multiple human tumor cell lines (IC50 = 1.25 nM in HeLa cells) and blocks metaphase/anaphase transition by suppression of microtubule dynamics (IC50 = 3.8 nM). Reduces spindle length by 29% and inhibits microtubule polymerization at micromolar concentrations.
Specifications

Chemical Identifiers

125317-39-7
1079.119
vinorelbine ditartrate
CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O
C53H66N4O20
CILBMBUYJCWATM-HJBYVSOESA-N
124080885

Specifications

Vinorelbine ditartrate
125317-39-7
vinorelbine ditartrate
CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O
124080885
>98%
50mg
C53H66N4O20
CILBMBUYJCWATM-HJBYVSOESA-N
1079.119
1079.1
Videos