Allsigns International™ Vinly Prohibition Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: SA2/SA.

Safety sign SA2/SA fire exit with runningman/directional arrow self adhesive vinyl right

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12309767

Quantity Price
1 £ 17.67 / Each
Estimated Shipment
02-03-2017
Add to basket


Description and Specification

Specification

Material Vinyl
Certifications/Compliance EEC 92/58/EEC
Message Fire exit with running man/right pointing arrow
Quantity 1Each
Color Green on White
Type Safety sign
Adhesive Type Self-adhesive
Dimensions (L x W) 600 x 200mm

Fire exit with running man/directional arrow self adhesive vinyl right pointing arrow green on white 600mm x 200mm Allsigns

Documentation

Technical documents