Learn More
Vinculin Monoclonal Antibody (7F9), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£149.00 - £406.00
Specifications
Clone | 7F9 |
---|---|
Concentration | 0.2 mg/mL |
Classification | Monoclonal |
Conjugate | eFluor 570 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558976
|
Affymetrix eBioscience
41-9777-82 |
100 μg |
£406.00
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15548976
|
Affymetrix eBioscience
41-9777-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody 7F9 (VIIF9) recognizes human, mouse, rat, and avian vinculin and its alternatively spliced isoform, metavinculin. Vinculin is a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Vinculin is involved in the anchoring of F-actin to the membrane and the regulation of E-cadherin expression. Vinculin binds to talin, paxillin, and alpha-actinin. Disregulation of vinculin alters cell adhesion, migration, and growth, which promotes cancer invasion and metastasis. Applications Reported: This 7F9 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry. Applications Tested: This 7F9 antibody has been tested by immunocytochemistry of formaldehyde-fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Vinculin Monoclonal antibody specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
7F9 | |
Monoclonal | |
Liquid | |
RUO | |
P18206, P85972, Q64727 | |
VCL | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
VCL |
0.2 mg/mL | |
eFluor 570 | |
Mouse | |
Human, Mouse, Rat | |
22330, 305679, 7414 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.