missing translation for 'onlineSavingsMsg'
Learn More

Vinculin, Biotin, clone: 7F9, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9777-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15567386

  • £69.00 / 25µg

Estimated Shipment
19-02-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody 7F9 (VIIF9) recognizes human, mouse, rat, and avian vinculin and its alternatively spliced isoform, metavinculin. Vinculin is a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Vinculin is involved in the anchoring of F-actin to the membrane and the regulation of E-cadherin expression. Vinculin binds to talin, paxillin, and alpha-actinin. Disregulation of vinculin alters cell adhesion, migration, and growth, which promotes cancer invasion and metastasis.Applications Reported: This 7F9 antibody has been reported for use in microscopy, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This 7F9 antibody has been tested by immunocytochemistry on formaldehyde-fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

vinculin
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P18206, P85972, Q64727
RP11-178G16.3, CMD1W, MVCL
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Murine, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
7F9
Biotin
VCL
Liquid
VCL
IgG1, kappa, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
22330, 305679, 7414
Videos
SDS
Documents

For Research Use Only.