missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin, Alexa Fluor 488, clone: 7F9, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 53-9777-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody 7F9 (VIIF9) recognizes human, mouse, rat, and avian vinculin and its alternatively spliced isoform, metavinculin. Vinculin is a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Vinculin is involved in the anchoring of F-actin to the membrane and the regulation of E-cadherin expression. Vinculin binds to talin, paxillin, and alpha-actinin. Disregulation of vinculin alters cell adhesion, migration, and growth, which promotes cancer invasion and metastasis.Applications Reported: This 7F9 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.Applications Tested: This 7F9 antibody has been tested by immunocytochemistry of formaldehyde-fixed and permeabilized cells and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
vinculin | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Q64727, P85972, P18206 | |
RP11-178G16.3, CMD1W, MVCL | |
Murine | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Rat |
Microscopy | |
7F9 | |
Alexa Fluor 488 | |
VCL | |
Conjugated | |
Vcl | |
IgG1, kappa | |
100μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
7414 |
For Research Use Only.