Learn More
Vincristine sulfate, Tocris Bioscience™
Disrupts microtubules
Brand:  RD Systems (Bio-Techne) 1257/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13488993
  • £147.15 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Anticancer agent; microtubule disrupter. Induces apoptosis in human lymphoma cells.
Specifications
Chemical Identifiers
2068-78-2
923.044
22-Oxovincaleukoblastine
C46H58N4O14S
AQTQHPDCURKLKT-DJOZZVTQSA-N
126963557
Specifications
Vincristine sulfate
>96%
22-Oxovincaleukoblastine
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
126963557
10mg
2068-78-2
C46H58N4O14S
AQTQHPDCURKLKT-DJOZZVTQSA-N
923.044
923.04
Videos