Learn More
Vinblastine sulfate, Tocris Bioscience™
Disrupts microtubules
Brand:  Tocris (Bio-Techne) 125650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13468993
  • £480.69 / 50mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Anticancer agent; microtubule disrupter. Induces apoptosis in cultured hepatocytes and human lymphoma cells. Shown to inhibit autophagosome maturation.
Specifications
Chemical Identifiers
143-67-9
909.061
vinblastine sulfate
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
C46H60N4O13S
KDQAABAKXDWYSZ-IEGOSMDXSA-N
131709225
Specifications
50mg
143-67-9
vinblastine sulfate
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
131709225
>95%
Vinblastine sulfate
C46H60N4O13S
KDQAABAKXDWYSZ-IEGOSMDXSA-N
909.061
909.06