missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Vimentin Rabbit Monoclonal Antibody
Vimentin, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
£128.00 - £520.00
Specifications
Antigen | Vimentin |
---|---|
Clone | SP20 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12643767
|
Epredia™
RM-9120-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12653767
|
Epredia™
RM-9120-S1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12633767
|
Epredia™
RM-9120-S |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12623767
|
Epredia™
RM-9120-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.Host Species: Rabbit
Clone: SP20
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding human vimentin
Molecular Weight: 57-60kDa
Positive Control: Sarcoma and melanoma
Cellular Localization: Cytoplasmic Clone SP20 is excellent for staining of formalin/paraffin with no special pretreatment.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Vimentin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG |
SP20 | |
Monoclonal | |
Rabbit | |
Cytoskeletal Protein | |
Recombinant protein encoding human vimentin | |
Primary |