missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ VIM (Human) ELISA Kit

Sandwich enzyme immunoassay for the quantitative measurement of human VIM

Brand:  Abnova™ KA4803.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16089085

  • £527.81 / 1 set


Estimated Shipment
18-06-2020
Add to basket

Description

Description

  • Used for the in vitro quantitative determination of human vimentin in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human vimentin
Specifications

Specifications

P08670
0.78 to 50 ng/mL
VIM (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human VIM.
7431
ELISA Kit
100 μL
Human
Colorimetric
Biotin
Quantification
Human vimentin microtiter plate, Plate sealers: Adhesive sheet, Washing Buffer (20x), Reconstitution buffer, Standard Protein, Standard/Sample Dilution Buffer, Secondary Antibody, Streptavidin HRP(X100), Secondary antibody/ Streptavidin HRP Dilution Buffer, Substrate (TMB), Stop Solution: 1 N solution of sulfuric acid (H2SO4)
For research use only (RUO)
Store the kit at 4°C.
Serum
Videos
Documents