Learn More
Vigabatrin, Tocris Bioscience™
GABA-T inhibitor
Brand:  Tocris (Bio-Techne) 080810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13428033
  • £119.07 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Selective GABA-T (transaminase) inhibitor. Anticonvulsant.
Specifications
Chemical Identifiers
60643-86-9
129.159
vigabatrin, sabril, vigabatrine, gamma-vinyl gaba, 4-amino-5-hexenoic acid, vigabatrina, vigabatrinum, gamma-vinyl-gaba, vigabatrine french, vigabatrinum latin
CHEBI:63638
C=CC(CCC(=O)O)N
C6H11NO2
PJDFLNIOAUIZSL-UHFFFAOYSA-N
5665
4-aminohex-5-enoic acid
Specifications
10mg
60643-86-9
vigabatrin, sabril, vigabatrine, gamma-vinyl gaba, 4-amino-5-hexenoic acid, vigabatrina, vigabatrinum, gamma-vinyl-gaba, vigabatrine french, vigabatrinum latin
C=CC(CCC(=O)O)N
129.159
CHEBI:63638
>98%
Vigabatrin
C6H11NO2
PJDFLNIOAUIZSL-UHFFFAOYSA-N
4-aminohex-5-enoic acid
5665
129.16