missing translation for 'onlineSavingsMsg'
Learn More

VICI Jour Chromatography™ PEEK Tubing

Brand:  VICI Jour Chromatography™ JR-T-4036-M5

Code : 16

Additional Details : Weight : 1.00000kg

Product Code. 13173573

  • £46.06 / 5m
Estimated Shipment: 24-04-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Tubing
Suitable for Lower Pressure
PTFE
Natural
1.58 mm
100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers