missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Jour Chromatography™ Striped PEEK Tubing, 1/16 in.
Color-coding identifies the ID of your PEEK tubing – each ID is coded with a different color.
Brand: VICI Jour Chromatography™ 724JRT6002M3
Code : 16
Additional Details : Weight : 1.00000kg
Description
- PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting
- Not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel
- Polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample
- Dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK
Specifications
Tubing | |
Maximum Chemical Resistance and Biocompatibility. | |
1.58 mm | |
100°C |
Orange | |
0.5 mm | |
PEEK |