missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Chemware™ PFA Vials with Tray
Saint Gobain Performance Plastics™ Chemware™ PFA Vials with Tray can be used with aggressive chemicals such as acids, bases, stains, chlorinated solvents and reagents.
Brand: Saint-Gobain D1069047
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Chemically resistant
- Extra sturdy construction for long life
- Includes polyethylene tray, 10 vials and 10 septa
- Ideal for trace metal analysis or any application where the use of glass can alter results
Specifications
18.3 mm | |
PFA | |
Polyethylene Tray and 10 PFA/Silicone Laminated Septa | |
Screw Cap | |
10 |
6.35 mm | |
7 mL | |
PFA Septa Style | |
36.2 mm |