Learn More
Saint-Gobain Silicone Versilic™ Cap
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Saint Gobain Performance Plastics™ Silicone Versilic™ Caps are particularly suited for all types of tissue culture work.
Brand: Saint-Gobain 26302420
Code : 02
Additional Details : Weight : 0.17600kg
Description
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifications
Silicone |